Learn More
Bioworld DMEM/Ham's F-12 (1:1) w/ HEPES, 1 L
Supplier:  Bioworld 306110231
Encompass_Preferred
Dublecco's Modified Eagles Medium/Nutrient Mixture Ham's F-12 (DMEM/Ham's F-12) was created after extensive study of Harry Eagle's first reports. Since then, studies have been underway to determine the media requirements for individual cell types. The medium found to be most satisfactory for these studies was a 1:1 mixture of Dublecco's Modified Eagle's Medium (DMEM) and Ham's F-12 Nutrient Mixture. To help compensate for the buffering capacity lost with the removal of serum, HEPES has been added in some modifications at a final concentration of 15mM. DMEM/Ham's F-12 (1:1 Mixture, powder), item #785060, contains L-glutamine, pyridoxine hydrochloride and 15mM HEPES buffer. Powder does not include sodium bicarbonate. Click here to see a DMEM/Ham's F12 1:1 Medium Components Table. Click here to see a Sodium Bicarbonate addition table for powdered media.
Catalog No. 50-254-067
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.