Learn More
Bioworld DMEM, High Glucose with L-glut & Pyridoxine HCL, w/o Sodium phosphate and Sodium pyruvate.(powder), 10 L
Supplier:  Bioworld 306110721
Encompass_Preferred
Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME), containing four-fold concentrations of the amino acids and vitamins found in BME, as well as additional supplementary components. The original formulation of DMEM contained 1000mg/L of glucose, but since has been raised in some modifications to 4500mg/L, giving the terms of Low Glucose or High Glucose. Several other modifications over the years, in addition to the alternative glucose levels, have broadened the spectrum of cell types supported by DMEM. DMEM (powder), High glucose with L-glutamine contains a pyridoxine hydrochloride supplement. DMEM does not include sodium phosphate, sodium bicarbonate or sodium pyruvate.
Catalog No. 50-254-172