Learn More
Bioworld DMEM, High Gluc w/o L-Glut, Phenol Red or Sodium Pyruvate, 500 mL
Supplier:  Bioworld 306110091
Encompass_Preferred
DMEM (liquid), High glucose without L-glutamine, item #785034, contains a pyridoxine hydrochloride supplement. Does not include phenol red or sodium pyruvate.Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME), containing four-fold concentrations of the amino acids and vitamins found in BME, as well as additional supplementary components. The original formulation of DMEM contained 1000mg/L of glucose, but since has been raised in some modifications to 4500mg/L, giving the terms of Low Glucose or High Glucose. Several other modifications over the years, in addition to the alternative glucose levels, have broadened the spectrum of cell types supported by DMEM.
Catalog No. 50-254-037