Learn More
Bioworld DMEM, High Gluc w/ Pyridoxine HCL w/o L-Glut, 1 L
Supplier:  Bioworld 306110121
Encompass_Preferred
Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME), containing four-fold concentrations of the amino acids and vitamins found in BME, as well as additional supplementary components. The original formulation of DMEM contained 1000mg/L of glucose, but since has been raised in some modifications to 4500mg/L, giving the terms of Low Glucose or High Glucose. Several other modifications over the years, in addition to the alternative glucose levels, have broadened the spectrum of cell types supported by DMEM. DMEM (powder), High glucose without L-glutamine, item #785031, contains a pyridoxine hydrochloride supplement. Does not include sodium pyruvate or sodium bicarbonate. Click here to see a Sodium Bicarbonate addition table for powdered media. Click here to see a DMEM High Glucose Medium Components Table.
Catalog No. 50-254-045