Learn More
Bioworld DMEM w/o Glucose w/ L-Glut and Pyridoxine HCL, 500 mL
Supplier:  Bioworld 306110201
Encompass_Preferred
Dulbecco's Modified Eagle's Medium (DMEM) is a modification of Basal Medium Eagle (BME), containing four-fold concentrations of the amino acids and vitamins found in BME, as well as additional supplementary components. The original formulation of DMEM contained 1000mg/L of glucose, but since has been raised in some modifications to 4500mg/L, giving the terms of Low Glucose or High Glucose. Several other modifications over the years, in addition to the alternative glucose levels, have broadened the spectrum of cell types supported by DMEM. DMEM (liquid), no glucose with L-glutamine, contains a pyridoxine hydrochloride supplement. Does not include sodium pyruvate or glucose.
Catalog No. 50-254-061
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.