Learn More
Cayman Chemical DMG-PEG 2000 1g
SDP
Supplier:  Cayman Chemical 339451 G
Encompass_Preferred
A lipid excipient; administration of FAH mRNA in DMG-PEG(2000)-containing lipid nanoparticles increases survival in a mouse model of HT-1
Catalog No. 50-294-6913