Learn More
Cayman Chemical DMG-PEG 2000 500mg
SDP
Supplier:  Cayman Chemical 33945500 MG
Encompass_Preferred
A lipid excipient; administration of FAH mRNA in DMG-PEG(2000)-containing lipid nanoparticles increases survival in a mouse model of HT-1
Catalog No. 50-295-1040