Learn More
Medchemexpress LLC Dmg-peg-mal (mw 2000) | 99.3% | 2000 (Average) | 50 MG
SDP
Supplier:  Medchemexpress LLC HY16703650MG
Encompass_Preferred
DMG-PEG-Mal (MW 2000) is a PEG lipid that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for use in drug delivery research.
  • Improves delivery efficiency of poorly soluble drugs
  • Enhances tissue specificity for poorly soluble drugs
  • Suitable for drug delivery research
Catalog No. 50-003-90547