Learn More
Apexbio Technology LLC Anti-GAPDH (7E4) Mouse Monoclonal Antibody 1ml
Supplier: Apexbio Technology LLC MA57811
Anti-GAPDH 7E4 Mouse Monoclonal Antibody is a monoclonal antibody targeting glyceraldehyde-3-phosphate dehydrogenase (GAPDH) a key enzyme in glycolysis It is designed to specifically recognize GAPDH protein which is ubiquitously expressed across various tissues and metabolic states Anti-GAPDH 7E4 Mouse Monoclonal Antibody exerts its biological activity primarily through antigen-antibody binding to epitopes on GAPDH conserved across species including Human Monkey Rat Mouse and Hamster Based on these properties Anti-GAPDH 7E4 Mouse Monoclonal Antibody holds research potential in applications such as protein expression assays most notably as an internal loading control in Western blot (WB) analysis