Learn More
Apexbio Technology LLC DMH-1, 25mg. Cas: 1206711-16-1 MFCD: MFCD18384963. Selective BMP ALK2 receptor
SDP
Supplier:  Apexbio Technology LLC B368625
Encompass_Preferred
DMH1 is a small molecule inhibitor of bone morphogenetic protein (BMP) signaling with IC50 value of 107.9nM against ALK2 [1].DMH1 is an analog of dorsomorphin. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5349