Learn More
Selleck Chemical LLC DMH1 10mg 1206711-16-1
SDP
Supplier:  Selleck Chemical LLC S714610MG
Encompass_Preferred
DMH1 is a selective BMP receptor inhibitor with IC50 of 107.9 nM for ALK2, exhibiting no inhibition on AMPK, ALK5, KDR (VEGFR-2) or PDGFR. DMH1 inhibits autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4456