Learn More
Medchemexpress LLC HY-129098 10mg , DMHCA CAS:79066-03-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12909810MG
Encompass_Preferred
HY-129098 10mg DMHCA CAS: 79066-03-8 DMHCA, a potent and selective LXR agonist, specifically activates the cholesterol efflux arm of the LXR pathway without stimulating triglyceride synthesis. DMHCA has anti-inflammatory effects and can be used for the research of cholesterol homeostasis diabetes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0677