Learn More
Cole-Parmer Fluke 287/FVF View Forms Combo Kit with 287
Supplier:  Cole-Parmer 2604892
Encompass
This industrial meter for demanding users features built-in logger records reading and time of day to catch intermittent problems. The data can be viewed on the meter in what's called, TrendCapture, or optional FlukeView software. The meter also features peak capture to measure transients as fast as 250 S.
Catalog No. 50-630-962