Learn More
Cole-Parmer Fluke 179 True RMS Multimeter with backlight and temperature
Supplier:  Cole-Parmer 2607901
Encompass
These multimeters all feature a 6000-count resolution, display and auto hold, frequency, capacitance, resistance, continuity, and diode measurement capability. Meters include easy battery exchange, closed case calibration through the front panel, and an ergonomic case with integrated protective holster. Models 177 and 179 feature True RMS voltage and current measurement capabilities, and provide digital display with analog bar graph and backlight. Model 77 IV also includes average responding AC measurements, and model 179 features temperature measurement capabilities. Available for sale in the United States and Canada only.
Catalog No. 50-993-337