Learn More
Apexbio Technology LLC DMNPE-caged ATP diammonium salt 10mg
SDP
Supplier:  Apexbio Technology LLC B581310
Encompass_Preferred
DMNPE-caged ATP diammonium salt is a photo-sensitive ATP derivative designed to enable precise spatial and temporal control of ATP availability It achieves this by chemical modification of ATP with a DMNPE moiety which transiently suppresses its biological activity DMNPE-caged ATP exerts its bioactivity via rapid photolytic cleavage of the DMNPE group upon ultraviolet illumination restoring functional ATP Based on these properties DMNPE-caged ATP diammonium salt is employed as a tool in biological research fields such as cellular energetics ATP-dependent signaling pathway studies enzymatic regulation investigations and functional assays of ion channels
Catalog No. 50-000-91287