Learn More
Selleck Chemical LLC DMOG 20mg 89464-63-1 Dimethyloxalylglycine
SDP
Supplier:  Selleck Chemical LLC S748320MG
Encompass_Preferred
DMOG (Dimethyloxalylglycine) is an antagonist of ?-ketoglutarate cofactor and inhibitor for HIF prolylhydroxylase, leading to stabilisation and accumulation of HIF-1? protein in the nucleus. DMOG enhances autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4833