Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMP 696 | 202578-52-7 | >97.0% | 25 MG
SDP

Supplier:  Medchemexpress LLC HY1213125MG

Encompass_Preferred

DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist supplied as a research reagent for preclinical studies of anxiety and depression. It is provided as a small-molecule powder intended for in vitro and in vivo experimentation.

  • Selective CRHR1 antagonist activity.
  • Suitable for in vitro and in vivo studies.
  • Provided as a 25 mg powder for dosing and formulation.
  • CAS registry number 202578-52-7.
  • Molecular formula C18H21Cl2N5O2; molecular weight 410.3 g/mol.

Catalog No. 50-002-33740


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.