Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMP 696 | 202578-52-7 | 98.7% | 1 ML
SDP

Supplier:  Medchemexpress LLC HY1213110MM 1ML

Encompass_Preferred

DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist supplied for research use. It is offered as a 10 mM solution in DMSO (1 mL) and as solid powder in multiple milligram sizes. CAS: 202578-52-7.

  • Selective CRHR1 antagonist for preclinical research.
  • Available as 10 mM solution in DMSO (1 mL) and as solid in mg quantities.
  • Reported purity approximately 98.7%.
  • Molecular weight 410.30 g/mol; formula C18H21Cl2N5O2.
  • Physical form: solid powder; solution provided ready for use in DMSO.
  • Intended for research use only.

Catalog No. 50-002-32311


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.