Learn More
Medchemexpress LLC DMP 777 | 157341-41-8 | 99.8% | C31H40N4O6 | 1 ML

Supplier: Medchemexpress LLC HY7595710MM/1ML
DMP 777 is a potent, selective, and orally active human leukocyte elastase (HLE) inhibitor. This compound is intended for research use only and exhibits high purity.
- Potent, selective, and orally active human leukocyte elastase (HLE) inhibitor.
- Exhibits high purity.
- Stored at -20°C under nitrogen.
- When in solvent, store at -80°C for 6 months or -20°C for 1 month.
- Available as a white to yellow solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.