Learn More
Cayman Chemical DMPE-MPEG 2000 100mg
SDP
Supplier:  Cayman Chemical 34649100 MG
Encompass_Preferred
An oligosaccharide comprised of nine a-14-linked glucose molecules has been used as a substrate to study the cleavage distribution and enzyme kinetics of B. licheniformis thermostable a-amylase as well as the enzyme kinetics of A. niger glucoamylase II
Catalog No. 50-294-9892