Learn More
Medchemexpress LLC DMPG sodium | 200880-40-6 | 99.2% | 688.85 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY22274
Encompass_Preferred
DMPG sodium is a biochemical reagent used as a liposome carrier. It is utilized for loading 6-shogaol, which exhibits anticancer activity.
  • Functions as a biochemical reagent
  • Acts as a liposome carrier
  • Used to load 6-shogaol
  • Supports anticancer activity through 6-shogaol loading
Catalog No. 50-004-02342