Learn More
AdipoGen Dimethylsulfoniopropionate (DMSP) . Hydrochloride
SDP
Supplier:  AdipoGen BVT0468M100
Encompass_Preferred
Dimethylsulfoniopropionate (DMSP) . hydrochloride, AdipoGen Life Sciences. Chemical. CAS: 4337-33-1. Formula: C5H10O2S . HCl. MW: 134.2 . 36.5. Synthetic. Naturally occurring organosulfur compound. Osmolyte, osmoprotectant. Antioxidant. Cryoprotectant. Neuroprotective agent. Neuritogenic. Acts against immune-deficient diseases (e.g. cancer). Important in geochemistry, ecology and algal/plant physiology. Metabolite produced primarily by marine phytoplankton and is the main precursor to the climatically important gas dimethylsulfide (DMS).
Catalog No. 50-114-6886