Learn More
Medchemexpress LLC DMT-2′Fluoro-dU Phosphoramidite | 146954-75-8 | 100.0% | C39H46FN4O8P | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY45409
Encompass_Preferred
DMT-2' Fluoro-dU Phosphoramidite is a uridine phosphoramidite with a fluorine in the 2' position. It can be used for nucleoside modification and synthesis of DNA.
  • Uridine phosphoramidite
  • Contains fluorine in the 2' position
  • Used for nucleoside modification
  • Used for synthesis of DNA
Catalog No. 50-003-58673