Learn More
Medchemexpress LLC DMT-2 -O-MOE-rABz ph 10mM 1mL | 251647-53-7 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY10401710MM 1ML
Encompass_Preferred
DMT-2 -O-MOE-rA(Bz) phosphoramidite is an adenine nucleoside analog DMT-2 -O-MOE-rA(Bz) phosphoramidite can be used in research on oligonucleotide synthesis[1
Catalog No. 50-002-42551