Learn More
Medchemexpress LLC DMT-2′O-MOE-rG(ib) phosphoramidite | 251647-55-9 | MFCD31692297 | 95.0% | 913.99 | C47H60N7O10P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10401410MM 1ML
Encompass_Preferred
DMT-2′O-MOE-rG(ib) Phosphoramidite is a protected guanosine phosphoramidite for incorporation of 2'-O-methoxyethyl-modified guanosine in solid-phase oligonucleotide synthesis. It is supplied as a solid or as a ready-to-use 10 mM solution in DMSO and is suitable for stereochemical synthesis of phosphorothioate oligonucleotides and standard automated or manual coupling workflows.
  • Provides 2'-O-MOE-modified guanosine monomer for oligonucleotide synthesis.
  • Available as solid or ready-to-use 10 mM solution in DMSO.
  • Supports stereochemical synthesis of phosphorothioate oligonucleotides.
  • High purity for reliable coupling efficiency and product consistency.
  • Compatible with automated and manual solid-phase synthesis protocols.
  • Molecular formula and weight provided for analytical confirmation.
Catalog No. 50-002-41847