Learn More
Medchemexpress LLC DMT-2'-OMe-D-ribitol phosphoramidite | 1470023-72-3 | 99.2% | 650.74 | C36H47N2O7P | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16088410MG
Encompass_Preferred
DMT-2'-OMe-D-ribitol phosphoramidite is an abasic phosphoramidite monomer used to introduce the abasic group Y34 into synthetic oligonucleotides. It is supplied for research use and provided with analytical documentation to support identity and purity.
  • Abasic phosphoramidite monomer for incorporation of abasic group Y34 into oligonucleotides.
  • High purity (99.2%) supporting reproducible synthesis.
  • Analytical documentation available: COA, SDS, HNMR, RP-HPLC, MS.
  • Molecular weight 650.74 and chemical formula C36H47N2O7P.
  • Supplied in multiple small package sizes suitable for method development and synthesis optimization.
Catalog No. 50-002-88567