Learn More
Medchemexpress LLC Dmt-locmec(bz) phosphoramidite | 206055-82-5 | 95.0% | C48H54N5O9P | 100 MG

Supplier: Medchemexpress LLC HY49000100MG
DMT-locMeC(bz) phosphoramidite is a phosphorite monomer that can be used in the synthesis of oligonucleotides. This product is for research use only.
- Chemical name: DMT-locMeC(bz) phosphoramidite
- Cas number: 206055-82-5
- Purity: 95.0%
- Chemical formula: C48H54N5O9P
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.