Learn More
TARGETMOL CHEMICALS INC DMT1 BLOCKER 1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T110635MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DMT1 blocker 1 is a blocker of divalent metal transporter 1 (DMT1) with IC50 of 0.64 uM which is expected to block the absorption of iron by intestinal cells in vivo. purity: 100%
Catalog No. 50-258-1582