Learn More
Medchemexpress LLC DMU-212 | 134029-62-2 | 99.3% | 300.35 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13797750MG
Encompass_Preferred
DMU-212 is a methylated derivative of Resveratrol (HY-16561), which possesses antimitotic, anti-proliferative, antioxidant, and apoptosis-promoting activities. It functions by inducing mitotic arrest through the induction of apoptosis and activation of ERK1/2 protein. DMU-212 is orally active and is intended solely for research use.
  • Antimitotic activity
  • Anti-proliferative activity
  • Antioxidant activity
  • Apoptosis promoting activities
  • Induces mitotic arrest via apoptosis induction and ERK1/2 protein activation
  • Orally active
Catalog No. 50-003-48893