Learn More
Selleck Chemical LLC DMX-5084 S0268-5MG
SDP
Supplier:  Selleck Chemical LLC S02685MG
Encompass_Preferred
DMX-5804 is a potent orally active and selective inhibitor of Mitogen-activated protein kinase kinase kinase kinase-4 (MAP4K4) with IC50 of 3 nM for human MAP4K4 DMX-5804 is more potent on human MAP4K4 with pIC50 of 8 55 than MINK1/MAP4K6 and TNIK/MAP4K7 with pIC50 of 8 18 and 7 96 respectively
Catalog No. 50-000-41374