Learn More
Selleck Chemical LLC DMXAA (Vadimezan) 100mg 117570-53-3 NSC 640488, ASA-404
SDP
Supplier:  Selleck Chemical LLC S1537100MG
Encompass_Preferred
DMXAA (Vadimezan) is a vascular disrupting agents (VDA) and competitive inhibitor of DT-diaphorase with Ki of 20 ?M and IC50 of 62.5 ?M in cell-free assays, respectively. DMXAA (Vadimezan) is also a STING agonist with potential antineoplastic activity. DMXAA (Vadimezan) potently induces IFN-? but relatively low TNF-? expression in vitro. DMXAA (Vadimezan) has antiviral activity. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-635-9