Learn More
Cayman Chemical DmyoInosItol1 3 4trIphosp 1mg
SDP
Supplier:  Cayman Chemical 609721 MG
Encompass_Preferred
A member of the InsP family of small, soluble second messengers; inhibits Ins(3,4,5,6)P4 kinase activity to increase the cellular level of Ins(3,4,5,6)P4
Catalog No. 50-289-7020