Learn More
Cayman Chemical DmyoInositol1 3 5triphosphate
SDP
Supplier:  Cayman Chemical 10007781100 UG
Encompass_Preferred
A synthetic, pure phosphoinositol for signal transduction research; 50-fold less potent than Ins(1,4,5)P3 at initiating calcium release when injected into Xenopus oocytes when tested as the meso compound
Catalog No. 50-288-9387