Learn More
Cayman Chemical DmyoInositol2 4 5triphosphate
SDP
Supplier:  Cayman Chemical 100077791 MG
Encompass_Preferred
A metabolically stable IP3 receptor agonist; a partial agonist at rat hepatic IP3 receptors, exhibiting 65% of the maximal calcium response obtained with Ins(1,4,5)P3
Catalog No. 50-288-8015