Learn More
Worthington Biochemical Corporation DNA CELLULOSE DBL STRND 5G
SDP
Supplier:  Worthington Biochemical Corporation LS01122
Encompass
Prepared by a method developed at Worthington in which native, double-stranded calf thymus DNA is covalently bound to cellulose. Suitable for the purification of many DNA binding proteins such as polymerases, transcription factors, and terminators, etc.
Catalog No. NC9867783