Learn More
Zymo Research Corporation DNA Clean & Concentrator-5 Kit (10 Preps) w/ Zymo-Spin™ I Columns (Uncapped)
SDP
Supplier:  Zymo Research Corporation D4003T
Encompass_Preferred
It can also be used for post-RT cDNA clean-up andpurification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield DNA that is free of salts and contaminants in just 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restrictiondigestion procedures.
Catalog No. 50-125-1376