Learn More
Medchemexpress LLC DNA, d(P-thio)(T-C-G-T-C-G-T-T-T-T-C-G-G-C-G-C-G-C-G-C-C-G) | 1254617-22-5 | DNA, d(P-thio)(T-C-G-T-C-G-T-T-T-T-C-G-G-C-G-C-G-C-G-C-C-G) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1507435MG
Encompass_Preferred
ODN 2395 is a C class oligodeoxynucleotide that functions as a vaccine adjuvant and a TLR9 agonist. It can be utilized in the research of immunological vaccines.
  • Functions as a vaccine adjuvant
  • TLR9 agonist
  • Used in immunological vaccine research
  • C class oligodeoxynucleotide
Catalog No. 50-003-75682