Learn More
Bioworld DNA Denaturing Solution, 500 mL
Supplier:  Bioworld 107500061
Encompass_Preferred
Denaturing Solution for use in Acrylamide Gel ElectrophoresisSpecificationContains: 0.5N NaOH and 1.5M NaCl
Catalog No. 50-196-820