Learn More
AMPLYUS LLC miniPCR bio miniPCR DNA Discovery System™ Pro
SDP
Supplier:  AMPLYUS LLC QP202016
Encompass_Preferred
Your professional biotech toolkit. miniPCR DNA Discovery System™ Pro includes a mini16X thermal cycler (portable and durable construction, heated lid, 16 x 0.2 ml PCR tube capacity, Bluetooth control from phone or laptop) paired with the GELATO™ electrophoresis and visualization system.
Catalog No. 50-223-7921