Learn More
ATCC Genomic DNA, Malassezia restricta (strain BS 7877) - 5 μg, dried (ATCC MYA-4611D-5)
Supplier:  ATCC MYA4611D5
Encompass
Genomic DNA isolated from Malassezia restricta strain BS 7877. This dried preparation contains 5 μg of high molecular weight genomic DNA stored in 1X TE buffer and is suitable for PCR, next-generation sequencing, and other molecular biology applications. Integrity and identity were verified by electrophoresis and sequencing, and functional activity was confirmed by PCR.
  • High molecular weight genomic DNA suitable for PCR and sequencing.
  • Mass: 5 μg.
  • Format: dried; stored in 1X TE buffer.
  • Quality control: no detectable RNA by electrophoresis.
  • Integrity verified by 1% agarose gel electrophoresis.
  • Functional activity confirmed by PCR amplification of ~1500 bp rRNA fragment.
  • Identity confirmed by ITS region sequencing and whole-genome sequencing.
  • Intended applications: next-generation sequencing and molecular biology workflows.
Catalog No. NC3355791