Learn More
ATCC Genomic DNA from Human Adenovirus 51 strain Bom, VR-1603D
Supplier: ATCC VR1603D
Genomic DNA isolated from human adenovirus 51 strain Bom. Total DNA was extracted from infected A549 cells and prepared using methods known to inactivate viruses. Supplied for laboratory research use only; store at -70°C or colder.
- Genomic DNA from human adenovirus 51 strain Bom.
- Extracted from infected A549 cells (total DNA).
- Prepared using methods known to inactivate viruses.
- Supplied as 100 μL per vial.
- Concentration not stated; contact manufacturer for lot-specific concentration.
- Store at -70°C or colder.
- Intended for laboratory research use only; not for therapeutic, diagnostic, or consumption use.
- Strain provenance: RIVM no. 89-13959.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.