Learn More
Sigma Aldrich Fine Chemicals Biosciences DNA Isolation Kit for Cell
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11814770001
Encompass
The DNA Isolation Kit for Cells and Tissues provides medium- and large-scale preparation of purified genomic DNA ranging in size from 50 to 150 kb. Remove contaminating RNA and proteins from a wide variety of biological specimens (mammalian tissue cultured cells yeast gram-negative bacteria or mouse tail).
Catalog No. 50-290-5732