Learn More
Biotium DNA Ligase 1(1A9), CF594 conjugate, 0.1mg/mL
Supplier:  Biotium BNC942177100
Encompass_Preferred
DNA Ligase I maintains the major DNA Ligase activity in proliferating cells by joining Okazaki fragments during lagging strand DNA replication. Human DNA Ligase I also has an essential role in DNA repair pathways, where it catalyzes the formation of phosphodiester bonds between adjacent 5' phosphoryl and 3' hydroxy termini at single breaks in duplex DNA molecules. In addition, DNA Ligase I plays a role in sealing nicks during excision repair. Similar to other DNA ligases, DNA Ligase I is built around a common catalytic core. Increased levels of DNA Ligase I are found in human tumors, as compared to benign tissues, as well as in peripheral blood lymphocytes. DNA Ligase I activity is altered in the chromosomal breakage deficit Bloom's syndrome (BS). Individuals with BS either have decreased levels of abnormally thermolabile DNA Ligase I or possess a dimeric form of this enzyme. Primary antibodies are available purified, or with a selection of fluorescent CF Dyes and other labels. C
Catalog No. 50-006-70514