Learn More
Cayman Chemical DNA MethyltrnsfRase 3a Mon 1ea
SDP
Supplier:  Cayman Chemical 134821 EA
Encompass_Preferred
Antigen: recombinant mouse His-tagged DNMT3a expressed in bacteria • Host: mouse, clone 64B814.1 • Isotype: IgG1κ • Cross Reactivity: (+) human and mouse DNMT3a • Application(s): WB, IF, and ICC • The DNMT3 family members DNMT3a and DNMT3b are strongly expressed in embryonic stem cells. Their expression is down regulated in differentiating cells and low in adult sematic tissue.
Catalog No. 50-295-5621