Learn More
Cayman Chemical DNA MethyltrnsfRase 3a Mon 1ea
SDP
Supplier:  Cayman Chemical 134841 EA
Encompass_Preferred
Antigen: DNMT3a expressed in bacteria The epitope was found to be located near the C-terminus (amino acids 705-908) • Host: mouse, clone 64B1446 • Isotype: IgG1κ • Cross Reactivity: (+) human and mouse DNMT3a • Application(s): ChIP, IF/ICC, IHC (paraffin), and WB • The DNMT3 family members DNMT3a and DNMT3b are strongly expressed in embryonic stem cells. Their expression is down regulated in differentiating cells and low in adult sematic tissue.
Catalog No. 50-295-5064