Learn More
STA PHARMACEUTICAL US LLC DNA O6-Ph-I amidite | 200 mg | CAS 133471-07-5 | MDL MFCD08061992
Supplier: STA PHARMACEUTICAL US LLC STAAM10672200MG
DNA O6-Ph-I amidite is a Amidite reagent (Subcategory: Inosine Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. SDS available for reference.
Specifications
- CAS: 133471-07-5
- MDL: MFCD08061992
- InChIKey: MVDGBHSFCSFDSA-FPRICFGYSA-N
- Molecular Weight: 830.922761998
- Molecular Formula: C46H51N6O7P
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(6-phenoxy-9H-purin-9-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=NC6=C5N=CN=C6OC7=CC=CC=C7)C(C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.