Learn More
Medchemexpress LLC Dna-pk-in-8 | 2823369-81-7 | 99.7% | 394.43 g·mol-1 | C19H22N8O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1465655MG
Encompass_Preferred
DNA-PK-IN-8 is a potent, selective, orally active inhibitor of DNA-dependent protein kinase used in preclinical research to probe DNA repair mechanisms, assess cancer cell response, and evaluate combination therapies. It shows subnanomolar potency and has demonstrated antiproliferative and tumor-growth suppression activity in model systems.
  • Potent inhibition of DNA-PK (IC50 0.8 nM).
  • High purity suitable for research applications.
  • Orally active in preclinical models.
  • Useful for DNA repair and oncology studies.
  • Available in small-quantity packages for laboratory use.
Catalog No. 50-002-23114