Learn More
STA PHARMACEUTICAL US LLC DNA S4-CNEt-U amidite | 200 mg | CAS 144303-74-2 | MDL MFCD08062020
Supplier: STA PHARMACEUTICAL US LLC STAAM10208200MG
DNA S4-CNEt-U amidite is a Amidite reagent (Subcategory: Uracil Series) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 144303-74-2
- MDL: MFCD08062020
- InChIKey: OZGHGLGGZJXMCQ-WVVVMPSKSA-N
- Molecular Weight: 799.923761998
- Molecular Formula: C42H50N5O7PS
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: (2R,3S,5R)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(4-((2-cyanoethyl)thio)-2-oxopyrimidin-1(2H)-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1C[C@@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C=CC(SCCC#N)=NC5=O)OCCC#N)C(C)C)C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.