Learn More
Teknova DNA Suspension Buffer, pH 8.0. DNase/RNase Tested, PCR Grade. 1mL Tubes, 50 Pack, Sterile. 6 pack.
SDP
Supplier:  Teknova 2T02216PK
Encompass
TE buffer is a salt solution that is commonly used to resuspend (or dilute) DNA and RNA. Composed of Tris base and EDTA, these molecules work together to maintain pH and prevent nucleic acids from degradation.
Catalog No. 50-274-5938