Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA PHARMACEUTICAL US LLC DNA T 3'-DMTr 5'-amidite | 10 g | CAS 134031-86-0 | MDL MFCD08706371

Supplier:  STA PHARMACEUTICAL US LLC STAAM10512MM25010GM

Encompass

DNA T 3'-DMTr 5'-amidite is a Amidite reagent (Subcategory: Reverse Amidite) sold by WuXi TIDES. Offered in 10 g. Store at -20 °C. SDS available for reference.

Specifications
- CAS: 134031-86-0
- MDL: MFCD08706371
- InChIKey: PYRVCWBKTIQJDJ-CEXSRUIHSA-N
- Molecular Weight: 744.825761998
- Molecular Formula: C40H49N4O8P
- Purity: ≥95%
- Container Type: 250 mL Glass (28-400)
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 179.7 g
- Commodity Code: 29349990
- Country Of Origin: China
- IUPAC: ((2R,3S,5R)-3-(bis(4-methoxyphenyl)(phenyl)methoxy)-5-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-2-yl)methyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OC[C@H]1O[C@H](C[C@@H]1OC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)N5C(NC(C(C)=C5)=O)=O)OCCC#N)C(C)C)C

Catalog No. 50-236-4454


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.